Learn More
Invitrogen™ Carbonic Anhydrase II Monoclonal Antibody (10E11)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549145
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Human CA2 shares 81.1% and 80.7% amino acid (aa) sequence identity with mouse and rat CA2, respectively. Positive Control - WB: rat heart tissue, rat kidney tissue, rat spleen tissue, rat liver tissue, mouse heart tissue, mouse kidney tissue, mouse spleen tissue, mouse liver tissue, human placenta tissue, human 293T whole cell. IHC: human gastric cancer tissue, human colon cancer tissue, mouse intestine tissue, rat intestine tissue. ICC/IF: HEPG2 cell. Flow: 293T cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
CA2 is one of several (at least 7) isozymes of carbonic anhydrase. Carbonic anhydrase catalyzes reversible hydration of carbon dioxide. Defects in this enzyme are associated with osteopetrosis and renal tubular acidosis.
Specifications
| Carbonic Anhydrase II | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P00918, P00920, P27139 | |
| CA2, Car2 | |
| E.coli-derived human CA2 recombinant protein (Position: S2-K260). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 10E11 | |
| Unconjugated | |
| CA2 | |
| AI131712; CA II; CA2; CAC; CAII; CA-II; Car2; Car-2; Carbonate dehydratase II; carbonic anhydrase 2; carbonic anhydrase B; carbonic anhydrase C; Carbonic anhydrase II; carbonic dehydratase; epididymis luminal protein 76; epididymis secretory protein Li 282; HEL-76; HEL-S-282; Ltw-5; Lvtw-5 | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 12349, 54231, 760 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.