missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase II/CA2 Rabbit anti-Mouse, Clone: 3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90677-50UL
This item is not returnable.
View return policy
Description
Carbonic Anhydrase II/CA2 Monoclonal specifically detects Carbonic Anhydrase II/CA2 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Carbonic Anhydrase II/CA2 | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 3 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:100-1:500 | |
| CAII, CA-IIEC 4.2.1.1, Car2, Carbonate dehydratase II, carbonic anhydrase 2, carbonic anhydrase B, Carbonic anhydrase C, carbonic anhydrase IICAC, carbonic dehydratase | |
| Recombinant Mouse Carbonic Anhydrase II/CA2 protein | |
| 50 μg | |
| Hypoxia, Lipid and Metabolism | |
| 760 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto