missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase I/CA1 Antibody (26E3), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26469-100ul
This item is not returnable.
View return policy
Description
Carbonic Anhydrase I/CA1 Monoclonal antibody specifically detects Carbonic Anhydrase I/CA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Specifications
| Carbonic Anhydrase I/CA1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 26E3 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200, ELISA, Immunohistochemistry 1:50-1:200 | |
| CA-I, Car1, Carbonate dehydratase I, carbonic anhydrase 1, Carbonic anhydrase B, carbonic anhydrase ICAB, carbonic dehydratase, EC 4.2.1.1 | |
| A synthesized peptide derived from Human Carbonic Anhydrase I/CA1 [UniProt P00915] | |
| 100 μL | |
| Hypoxia, Lipid and Metabolism | |
| 759 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction