Learn More
Restek Carbon WR/PDMS SPME Fiber (Carbon W Range) Fiber thik 95um Fiber L 10mm 23 g needle Dark Blue
Brand:  Restek 27479.1
Encompass_Preferred
Carbon WR/PDMS SPME Fiber (Carbon W Range) Fiber thik 95um Fiber L 10mm 23 g needle Dark Blue
Product Code. 16688626
  • £235.95 / Each