missing translation for 'onlineSavingsMsg'
Learn More

Carbon Tetrabromide, TRC

Product Code. 30365192
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30365192 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30365192 Supplier Toronto Research Chemicals Supplier No. TRCC1769005G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrabromomethane
CAS 558-13-4
Molecular Formula C Br4
Quantity 5 g
Synonym Tetrabromomethane, Methane, tetrabromo-, Carbon tetrabromide (8CI), Tetrabromomethane, Carbon bromide (CBr4), Methane tetrabromide, NSC 6179
InChI Formula InChI=1S/CBr4/c2-1(3,4)5
SMILES BrC(Br)(Br)Br
IUPAC Name tetrabromomethane
Molecular Weight (g/mol) 331.63
Formula Weight 327.6734
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.