missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, Resulfurized, AISI 1141, G11410, 100g, ARMI∣MBH
Click to view available options
Quantity:
100 g
Unit Size:
Each
Description
Carbon Steel, Resulfurized, AISI 1141, G11410, 100g
Specifications
Specifications
| Chemical Name or Material | Calcium/Boron/Titanium/Aluminium/Vanadium/Oxygen/Arsenic/Cobalt/Nitrogen/Tin/Phosphorus/Molybdenum/Niobium/Nickel/Chromium/Copper/Silicon/Carbon/Manganese/Cadmium/Zinc/Selenium/Magnesium/Tantalum/Bismuth/Lead/Antimony/Zirconium/Tungsten |
| CAS | 7439-89-6 |
| Quantity | 100 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Calcium 0.001 %/Boron 0.0013 %/Titanium 0.0015 %/Aluminium 0.002 %/Vanadium 0.0029 %/Oxygen 0.003 %/Arsenic 0.007 %/Cobalt 0.01 %/Nitrogen 0.01 %/Tin 0.0112 %/Phosphorus 0.0121 %/Molybdenum 0.026 %/Niobium 0.027 %/Nickel 0.081 %/Chromium 0.123 %/Copper 0.23 %/Silicon 0.27 %/Carbon 0.384 %/Manganese 1.46 %/Cadmium (<0.001) %/Zinc (<0.003) %/Selenium (<0.005) %/Magnesium (<0.006) %/Tantalum (<0.008) %/Bismuth (<0.02) %/Lead (0.002) %/Antimony (0.003) %/Zirconium (0.003) %/Tungsten (0.009) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction