missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, Resulfurized, AISI 1117, G11170, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Carbon Steel, Resulfurized, AISI 1117, G11170, Ø40x20mm
Specifications
Specifications
| Chemical Name or Material | Lead/Aluminium/Arsenic/Tin/Nitrogen/Phosphorus/Molybdenum/Nickel/Copper/Chromium/Silicon/Carbon/Manganese |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Lead 0.0011 %/Aluminium 0.0023 %/Arsenic 0.0044 %/Tin 0.011 %/Nitrogen 0.0112 %/Phosphorus 0.0133 %/Molybdenum 0.0317 %/Nickel 0.0877 %/Copper 0.1101 %/Chromium 0.1126 %/Silicon 0.151 %/Carbon 0.154 %/Manganese 1.129 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction