missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, Residuals, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Carbon Steel, Residuals, Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Tin/Titanium/Phosphorus/Tungsten/Vanadium/Arsenic/Carbon/Molybdenum/Aluminium/Copper/Cobalt/Chromium/Silicon/Nickel/Manganese |
| CAS | 7439-89-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Tin 0.018 %/Titanium 0.018 %/Phosphorus 0.025 %/Tungsten 0.035 %/Vanadium 0.069 %/Arsenic 0.085 %/Carbon 0.175 %/Molybdenum 0.19 %/Aluminium 0.195 %/Copper 0.25 %/Cobalt 0.435 %/Chromium 0.455 %/Silicon 0.47 %/Nickel 0.475 %/Manganese 1.37 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction