missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, Rephosphorized and Resulfurized, AISI 12L14, G12144, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Carbon Steel, Rephosphorized and Resulfurized, AISI 12L14, G12144, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Titanium/Niobium/Zirconium/Boron/Vanadium/Aluminium/Arsenic/Tin/Silicon/Nitrogen/Molybdenum/Copper/Oxygen/Nickel/Chromium/Phosphorus/Carbon/Lead/Manganese/Antimony/Cobalt/Tungsten |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Titanium 0.0009 %/Niobium 0.001 %/Zirconium 0.001 %/Boron 0.0011 %/Vanadium 0.002 %/Aluminium 0.0021 %/Arsenic 0.003 %/Tin 0.003 %/Silicon 0.004 %/Nitrogen 0.0049 %/Molybdenum 0.01 %/Copper 0.016 %/Oxygen 0.016 %/Nickel 0.019 %/Chromium 0.055 %/Phosphorus 0.078 %/Carbon 0.079 %/Lead 0.18 %/Manganese 1.06 %/Antimony (0.001) %/Cobalt (0.002) %/Tungsten (0.002) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction