Learn More
Swann-Morton™ Carbon Steel Non-sterile Scalpel Blades
Use Swann Morton™ Carbon Steel Non-sterile Scalpel Blades for precise cutting in the operating room.
Marke: Swann-Morton™ 0104
Code : 03
Weitere Details : Weight : 0.07896kg
Beschreibung
Use Swann Morton™ Carbon Steel Non-sterile Scalpel Blades for precise cutting in the operating room.
- Carbon steel to BS EN 27740
- Handles and blades sold separately
Spezifikation
Carbon Steel | |
Suture Cutter but also for Arteriotomies, Parotid Surgeries | |
Non-sterile |
Scalpel Blade | |
# 12 | |
10/Pk. |