missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, AISI 1045, G10450, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Carbon Steel, AISI 1045, G10450, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Titanium/Niobium/Tungsten/Aluminium/Arsenic/Cobalt/Tin/Nitrogen/Phosphorus/Molybdenum/Vanadium/Nickel/Chromium/Silicon/Copper/Carbon/Manganese/Magnesium/Lead/Tantalum/Boron/Calcium/Antimony/Zinc/Oxygen |
| CAS | 7439-89-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Titanium 0.00013 %/Niobium 0.001 %/Tungsten 0.003 %/Aluminium 0.004 %/Arsenic 0.005 %/Cobalt 0.007 %/Tin 0.0079 %/Nitrogen 0.009 %/Phosphorus 0.0103 %/Molybdenum 0.0217 %/Vanadium 0.0244 %/Nickel 0.097 %/Chromium 0.109 %/Silicon 0.225 %/Copper 0.232 %/Carbon 0.453 %/Manganese 0.749 %/Magnesium (<0.001) %/Lead (<0.002) %/Tantalum (<0.005) %/Boron (0.0002) %/Calcium (0.001) %/Antimony (0.0019) %/Zinc (0.002) %/Oxygen (0.004) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction