missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbon Steel, AISI 1018, G10180, ARMI∣MBH
Click to view available options
Thickness (Metric):
3 mm
19 mm
Unit Size:
Each
Description
Carbon Steel, AISI 1018, G10180
Specifications
Specifications
| Chemical Name or Material | Aluminium, Arsenic, Boron, Carbon, Chromium, Cobalt, Copper, Manganese, Molybdenum, Nickel, Niobium, Nitrogen, Oxygen, Phosphorus, Silicon, Sulfur, Tin, Titanium, Tungsten, Vanadium |
| Diameter (Metric) Outside | 38 mm |
| Form | Disc |
| Thickness (Metric) | 19 mm |
| Concentration or Composition (by Analyte or Components) | Titanium: 9 ppm, Boron: 0.0006%, Vanadium: 0.002%, Arsenic: 0.004%, Niobium: 0.004%, Oxygen: 0.004%, Tungsten: 0.004%, Tin: 0.0048%, Cobalt: 0.0052%, Nitrogen: 0.0059%, Phosphorus: 0.008%, Molybdenum: 0.0126%, Aluminium: 0.021%, Sulfur: 0.025%, Nickel: 0.034%, Chromium: 0.061%, Copper: 0.076%, Carbon: 0.172%, Silicon: 0.26%, Manganese: 0.65% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction