missing translation for 'onlineSavingsMsg'
Learn More

Carbon(isothiocyanatidic)acid Ethyl Ester, TRC

Product Code. 30358008
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30358008 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30358008 Supplier Toronto Research Chemicals Supplier No. TRCC17665010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbon(isothiocyanatidic)acid, Ethyl Ester
CAS 16182-04-0
Molecular Formula C4 H5 N O2 S
Quantity 10 g
Synonym Carbon(isothiocyanatidic) acid, ethyl ester, Carbonic acid, ethyl ester, anhydride with isothiocyanic acid (7CI), Carbonic acid, monoanhydride with isothiocyanic acid, ethyl ester (8CI), Carbethoxyisothiocyanate, Carbonic acid ethyl ester isothiocyanate, Ethoxycarbonyl isothiocyanate, Ethoxycarbonyl mustard oil, Ethyl carbonisothiocyanatidate, Ethyl isothiocyanatocarbonate, Ethyl isothiocyanatoformate, NSC 158480
InChI Formula InChI=1S/C4H5NO2S/c1-2-7-4(6)5-3-8/h2H2,1H3
SMILES CCOC(=O)N=C=S
IUPAC Name ethyl N-(sulfanylidenemethylidene)carbamate
Molecular Weight (g/mol) 131.15
Formula Weight 131.0041
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.