missing translation for 'onlineSavingsMsg'
Learn More

Carbon felt, 3.18mm (0.125in) thick, 99.0%, Thermo Scientific Chemicals

Product Code. p-7122659
Click to view available options
Quantity:
10 x 10 cm
25 x 25 cm
Unit Size:
Each
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 11369258

Brand: Thermo Scientific Alfa Aesar 043199.RR

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Typical thermal conductivity of 1.75 BTU in/hr/ft²/°F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00133992

Specifications

Assay Percent Range 99.0%
Quantity 10 x 10 cm
Chemical Name or Material Carbon felt, Processed at 1400°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.