missing translation for 'onlineSavingsMsg'
Learn More

Carbon disulfide, CHROMASOLV™, for HPLC, ≥99.9%, Solstice

Product Code. 15602870
Change view
Click to view available options
Quantity:
100 mL
1 L
4 x 2 L
6 x 1 L
Packaging:
Glass Bottle
Unit Size:
1 L
100mL
Pack of 4
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15602870 6 x 1 L Glass Bottle Pack of 6
15672860 100 mL Glass Bottle 100mL
15682860 1 L Glass Bottle 1 L
15692860 4 x 2 L Glass Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15602870 Supplier Honeywell-Chromasolv Supplier No. 2706606X1L
  • ONLINE EXCLUSIVE PRICE
    £643.00
    £538.18 / Pack of 6
    Save £104.82
    16% Off
View all promotions
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHROMASOLV™, for HPLC,≥99.9%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-15-0
Molecular Formula CS2
Molecular Weight (g/mol) 76.131
MDL Number MFCD00011321
InChI Key QGJOPFRUJISHPQ-UHFFFAOYSA-N
Synonym carbon disulfide, carbon disulphide, carbon bisulfide, dithiocarbonic anhydride, carbon bisulphide, schwefelkohlenstoff, weeviltox, wegla dwusiarczek, solfuro di carbonio, carbondisulphide
PubChem CID 6348
ChEBI CHEBI:23012
IUPAC Name methanedithione
SMILES C(=S)=S

Specifications

Melting Point -112°C to -111°C (literature)
Absorbance max. 0.50 at 388 nm, max. 1.0 at 386 nm, max. 0.01 at 750 nm, max. 0.01 500 nm, max. 0.05 at 410 nm, max. 0.10 at 403 nm, max. 0.25 at 394 nm
CAS Max % 100.0000%
Density 1.266 g/mL at 25 °C(lit.)
Quantity 6 x 1 L
Flash Point −30°C
Packaging Glass Bottle
Linear Formula CS2
UN Number UN1131
Beilstein 1098293
Boiling Point 46°C
Formula Weight 76.14
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material Carbon disulfide, CHROMASOLV™, for HPLC, ≥99.9%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.