missing translation for 'onlineSavingsMsg'
Learn More

Carbon black, acetylene, 50% compressed, 99.9+%, Thermo Scientific Chemicals

Form:S.A. 75m2/g, Bulk density 80-120g/L

£43.50 - £279.00

Chemical Identifiers

CAS 1333-86-4
Molecular Formula C
Molecular Weight (g/mol) 12.01
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11354687
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039724.30
250 g
£43.50
250g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11334687
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039724.A1
1 kg
£110.00
1kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11344687
View Documents Promotion Details
Thermo Scientific Alfa Aesar
039724.A5
3 kg
£279.00
3kg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1333-86-4
12.01
OKTJSMMVPCPJKN-UHFFFAOYSA-N
C
MFCD00133992

Specifications

1333-86-4
Bulk density 80 to 120g/L
C
UN1325
12.01
Carbon black, acetylene, 50% compressed
Black
Odorless
MFCD00133992
OKTJSMMVPCPJKN-UHFFFAOYSA-N
≥99.9%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H351;
Flammable solid.
Suspected of causing cancer.

P201-P202-P281-P308+P313-P501c

H351

missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 215-609-9

missing translation for 'rtecsNumber' : FF5800000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only