missing translation for 'onlineSavingsMsg'
Learn More

Carbon (Amorphous) (13C, 99 atom %), TRC

Product Code. 17972135
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17972135 50 mg 50mg
17924655 10 mg 10mg
17941815 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17972135 Supplier Toronto Research Chemicals Supplier No. TRCC78016050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14762-74-4
Formula Weight 13.0034
InChI Formula InChI=1S/C/i1+1
IUPAC Name carbon-13
Molecular Formula 13C
Molecular Weight (g/mol) 13
SMILES [13C]
Chemical Name or Material Carbon-13

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.