missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carbol Fuchsin, pure

Product Code. 10723285 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10723285

Brand: Thermo Scientific Chemicals 195791000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4197-24-4 | C21H22ClN3 | 351.878 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4197-24-4
Molecular Formula C21H22ClN3
Molecular Weight (g/mol) 351.878
MDL Number MFCD00143923
InChI Key HZLHRDBTVSZCBS-GHTYLULLSA-N
PubChem CID 91997463
IUPAC Name 4-[(Z)-(4-aminophenyl)-(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]-2-methylaniline;hydron;chloride
SMILES [H+].CC1=CC(=C(C2=CC=C(C=C2)N)C3=CC(=C(C=C3)N)C)C=CC1=N.[Cl-]

Specifications

Infrared Spectrum Authentic
Lambda Max (in water) about 542nm
Grade Pure
Packaging Glass bottle
Suitability (for microscopy) passes
UV Spectrometry Complies
Color Green
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material Carbol Fuchsin, A mixture of Basic Fuchsin and phenol.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.