missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbohydrazide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
Carbohydrazide is widely utilized as a curing agent for epoxide-type resins and as an oxygen scrubber in boiler systems. It is applied in photography to stabilize color developers that produce images of the azo-methine and azine classes. It is used to develop ammunition propellants and stabilize soaps. It is also a useful reagent in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Carbohydrazide is widely utilized as a curing agent for epoxide-type resins and as an oxygen scrubber in boiler systems. It is applied in photography to stabilize color developers that produce images of the azo-methine and azine classes. It is used to develop ammunition propellants and stabilize soaps. It is also a useful reagent in organic synthesis.
Solubility
Soluble in water.
Notes
Store in cool place. Incompatible with strong oxidizing agents, strong acids, copper, zinc, nickel, lead and brass.
Carbohydrazide is widely utilized as a curing agent for epoxide-type resins and as an oxygen scrubber in boiler systems. It is applied in photography to stabilize color developers that produce images of the azo-methine and azine classes. It is used to develop ammunition propellants and stabilize soaps. It is also a useful reagent in organic synthesis.
Solubility
Soluble in water.
Notes
Store in cool place. Incompatible with strong oxidizing agents, strong acids, copper, zinc, nickel, lead and brass.
Specifications
Melting Point | 153°C to 158°C |
Linear Formula | (H2NNH)2C=O |
Quantity | 500 g |
UN Number | UN3077 |
Merck Index | 14,1804 |
Solubility Information | Soluble in water. |
Formula Weight | 90.09 |
Percent Purity | 97% |
Chemical Name or Material | Carbohydrazide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'