missing translation for 'onlineSavingsMsg'
Learn More

Carbohydrazide, 97%

Product Code. 11459053
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459053 25 g 25g
11469053 100 g 100g
11479053 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459053 Supplier Thermo Scientific Alfa Aesar Supplier No. A11145.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 497-18-7 | CH6N4O | 90.09 g/mol

Carbohydrazide is widely utilized as a curing agent for epoxide-type resins and as an oxygen scrubber in boiler systems. It is applied in photography to stabilize color developers that produce images of the azo-methine and azine classes. It is used to develop ammunition propellants and stabilize soaps. It is also a useful reagent in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbohydrazide is widely utilized as a curing agent for epoxide-type resins and as an oxygen scrubber in boiler systems. It is applied in photography to stabilize color developers that produce images of the azo-methine and azine classes. It is used to develop ammunition propellants and stabilize soaps. It is also a useful reagent in organic synthesis.

Solubility
Soluble in water.

Notes
Store in cool place. Incompatible with strong oxidizing agents, strong acids, copper, zinc, nickel, lead and brass.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-18-7
Molecular Formula CH6N4O
Molecular Weight (g/mol) 90.09
MDL Number MFCD00007591
InChI Key XEVRDFDBXJMZFG-UHFFFAOYSA-N
Synonym carbohydrazide, carbonic dihydrazide, carbazide, carbonohydrazide, carbodihydrazide, hydrazine, carbonyldi, hydrazine, carbonylbis, carbazic acid, hydrazide, semicarbazide, 4-amino, carbonic acid, dihydrazide
PubChem CID 73948
ChEBI CHEBI:61308
IUPAC Name 1,3-diaminourea
SMILES NNC(=O)NN

Specifications

Melting Point 153°C to 158°C
Linear Formula (H2NNH)2C=O
Quantity 25 g
UN Number UN3077
Merck Index 14,1804
Solubility Information Soluble in water.
Formula Weight 90.09
Percent Purity 97%
Chemical Name or Material Carbohydrazide
compliance-icons compliance-icons
Product Identifier
  • Carbohydrazide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: Carbonic dihydrazide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.