missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbohydrate Sulfotransferase 6/CHST6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32912
This item is not returnable.
View return policy
Description
Carbohydrate Sulfotransferase 6/CHST6 Polyclonal specifically detects Carbohydrate Sulfotransferase 6/CHST6 in Human samples. It is validated for Western Blot.
Specifications
| Carbohydrate Sulfotransferase 6/CHST6 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 6, carbohydrate sulfotransferase 6, C-GlcNAc6ST, corneal N-acetylglucosamine 6-sulfotransferase, Corneal N-acetylglucosamine-6-O-sulfotransferase, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.17, EC 2.8.2.21, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-beta, glcNAc6ST-5, Gn6st-5, GST4-beta, hCGn6ST, MCDC1, N-acetylglucosamine 6-O-sulfotransferase 5 | |
| Rabbit | |
| 44 kDa | |
| 100 μL | |
| Primary | |
| Human. Predicted cross-reactivity based on sequence identity: Bovine (92%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9GZX3 | |
| CHST6 | |
| Recombinant protein encompassing a sequence within the center region of human CHST5/6. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 4166 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction