missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbohydrate Sulfotransferase 6/CHST6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | Carbohydrate Sulfotransferase 6/CHST6 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
Carbohydrate Sulfotransferase 6/CHST6 Polyclonal specifically detects Carbohydrate Sulfotransferase 6/CHST6 in Human samples. It is validated for Western Blot.Specifications
| Carbohydrate Sulfotransferase 6/CHST6 | |
| Polyclonal | |
| Rabbit | |
| Q9GZX3 | |
| 4166 | |
| Synthetic peptides corresponding to CHST6(carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 6) The peptide sequence was selected from the middle region of CHST6. Peptide sequence YAFTGLSLTPQLEAWIHNITHGSGPGARREAFKTSSRNALNVSQAWRHAL. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 6, carbohydrate sulfotransferase 6, C-GlcNAc6ST, corneal N-acetylglucosamine 6-sulfotransferase, Corneal N-acetylglucosamine-6-O-sulfotransferase, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.17, EC 2.8.2.21, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 4-beta, glcNAc6ST-5, Gn6st-5, GST4-beta, hCGn6ST, MCDC1, N-acetylglucosamine 6-O-sulfotransferase 5 | |
| CHST6 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title