missing translation for 'onlineSavingsMsg'
Learn More

Carbohydrate Sulfotransferase 4/CHST4 Antibody (4D7), Novus Biologicals™

Product Code. 18377469 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377469 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377469 Supplier Novus Biologicals Supplier No. H00010164M10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Carbohydrate Sulfotransferase 4/CHST4 Monoclonal antibody specifically detects Carbohydrate Sulfotransferase 4/CHST4 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen Carbohydrate Sulfotransferase 4/CHST4
Applications ELISA
Classification Monoclonal
Clone 4D7
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_005760
Gene Alias carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 4, EC 2.8.2, EC 2.8.2.-, EC 2.8.2.17, EC 2.8.2.21, galactose/N-acetylglucosamine/N-acetylgalactosamine 6-O-sulfotransferase 3, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 3, glcNAc6ST-2, GlcNAc6ST2, Gn6st-2, GST3, GST-3, HEC-GlcNAc6ST, HECGLCNAC6ST, HEC-GLCNAC-6-ST, High endothelial cells N-acetylglucosamine 6-O-sulfotransferase, L-selectin ligand sulfotransferase, LSSTcarbohydrate sulfotransferase 4, N-acetylglucosamine 6-O-sulfotransferase 2
Host Species Mouse
Immunogen CHST4 (NP_005760, 221 a.a. ~ 320 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. LMIDSRIVMGQHEQKLKKEDQPYYVMQVICQSQLEIYKTIQSLPKALQERYLLVRYEDLARAPVAQTSRMYEFVGLEFLPHLQTWVHNITRGKGMGDHAF
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10164
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.