missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbohydrate Sulfotransferase 3/CHST3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98311-100ul
This item is not returnable.
View return policy
Description
Carbohydrate Sulfotransferase 3/CHST3 Polyclonal antibody specifically detects Carbohydrate Sulfotransferase 3/CHST3 in Mouse samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| Carbohydrate Sulfotransferase 3/CHST3 | |
| Polyclonal | |
| ELISA 1:5000-1:10000, Immunohistochemistry-Paraffin 1:250-1:1000 | |
| C6ST-1, C6ST1HSD, C6STEC 2.8.2.17, carbohydrate (chondroitin 6) sulfotransferase 3, carbohydrate sulfotransferase 3, Chondroitin 6-O-sulfotransferase 1, Chondroitin 6-sulfotransferase, EC 2.8.2, Galactose/N-acetylglucosamine/N-acetylglucosamine 6-O-sulfotransferase 0, GST-0 | |
| Produced in rabbits immunized with purified, recombinant Mouse Carbohydrate Sulfotransferase 3/CHST3 (Accession#: NP_058083.2; Glu39-Thr472) | |
| 100 μg | |
| Signal Transduction | |
| 9469 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur