missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbohydrate Sulfotransferase 15/CHST15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-59012
This item is not returnable.
View return policy
Description
Carbohydrate Sulfotransferase 15/CHST15 Polyclonal specifically detects Carbohydrate Sulfotransferase 15/CHST15 in Human samples. It is validated for Western Blot.
Specifications
| Carbohydrate Sulfotransferase 15/CHST15 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| B cell RAG associated protein (GALNAC4S-6ST), B-cell RAG-associated gene protein, BRAGKIAA0598RP11-47G11.1, carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15, carbohydrate sulfotransferase 15, DKFZp781H1369, EC 2.8.2.33, GalNAc4S-6ST, GALNAC4S6ST, hBRAG, MGC34346, N-acetylgalactosamine 4-sulfate 6-O-sulfotransferase | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51363 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q7LFX5 | |
| CHST15 | |
| Synthetic peptides corresponding to GALNAC4S-6ST(B cell RAG associated protein) The peptide sequence was selected from the middle region of GALNAC4S-6ST. Peptide sequence YDNSTDGEPPFLTQDFIHAFQPNARLIVMLRDPVERLYSDYLYFASSNKS. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Canine: 100%; Rat: 100%; Mouse: 100%; Human: 100%; Bovine: 92%; Chicken: 78%; Green puffer: 78%;. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction