missing translation for 'onlineSavingsMsg'
Learn More

Carbofuran-3-hydroxy, Fluka™

Product Code. 16242862 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 16242862

Brand: Honeywell-Fluka R164810MG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Analytical standard, reference material

Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials  and organic reference materials.

  • Produced according to ISO 9001
  • Suitable for a wide range of industries and applications
  • Detailed Certificate of Analysis included with every delivery
  • Expert Technical Support
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbofuran-3-hydroxy
Packaging Glass Bottle
CAS 16655-82-6
Color White
Melting Point 138°C to 141°C
Synonym 3-Hydroxycarbofuran; Methylcarbamic acid 2,3-dihydro-3-hydroxy-2,2-dimethyl-7-benzofuranyl ester
Industry Type Environmental Analysis
Molecular Weight (g/mol) 237.25
Quantity 10 mg
Grade Analytical Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.