missing translation for 'onlineSavingsMsg'
Learn More

Carbocisteine Diisopropyl Ester, Mikromol™

Product Code. 30050317
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050317 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050317 Supplier Mikromol Supplier No. MM0236.09

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Carbocisteine Diisopropyl Ester
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbocisteine Diisopropyl Ester
CAS 792135-12-7
Analyte or Component Names Carbocisteine Diisopropyl Ester
Shipping Condition Ice pack (-20°C)
Molecular Formula C11 H21 N O4 S
Certifications/Compliance ISO 17025
Synonym S-[2-(1-Methylethoxy)-2-oxoethyl]-L-cysteine 1-Methylethyl Ester
InChI Formula InChI=1S/C11H21NO4S/c1-7(2)15-10(13)6-17-5-9(12)11(14)16-8(3)4/h7-9H,5-6,12H2,1-4H3/t9-/m0/s1
SMILES CC(C)OC(=O)CSC[C@H](N)C(=O)OC(C)C
IUPAC Name propan-2-yl (2R)-2-amino-3-(2-oxo-2-propan-2-yloxyethyl)sulfanylpropanoate
Molecular Weight (g/mol) 263.35
Quantity 100 mg
Formula Weight 263.1191 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.