missing translation for 'onlineSavingsMsg'
Learn More

Carbidopa Monohydrate, TRC

Product Code. 30363229
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30363229 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30363229 Supplier Toronto Research Chemicals Supplier No. TRCC176118100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbidopa
CAS 38821-49-7
Molecular Formula C10 H14 N2 O4 . H2 O
Quantity 100 mg
Synonym Carbidopa, (2S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoic acid monohydrate, (S)-Carbidopa monohydrate, Benzenepropanoic acid, α-hydrazino-3,4-dihydroxy-α-methyl-, monohydrate, (S)-, Benzenepropanoic acid, α-hydrazino-3,4-dihydroxy-α-methyl-, monohydrate, (αS)- (9CI), Carbidopa monohydrate
InChI Formula InChI=1S/C10H14N2O4.H2O/c1-10(12-11,9(15)16)5-6-2-3-7(13)8(14)4-6;/h2-4,12-14H,5,11H2,1H3,(H,15,16);1H2/t10-;/m0./s1
SMILES O.C[C@@](Cc1ccc(O)c(O)c1)(NN)C(=O)O
IUPAC Name (2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid;hydrate
Molecular Weight (g/mol) 244.24
Formula Weight 244.1059
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.