missing translation for 'onlineSavingsMsg'
Learn More

Carbidopa, Mikromol™

Product Code. 30050769
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050769 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30050769 Supplier Mikromol Supplier No. MM1501.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Carbidopa
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbidopa
CAS 38821-49-7
Analyte or Component Names Carbidopa
Shipping Condition Room Temperature
Molecular Formula C10 H14 N2 O4 . H2 O
Certifications/Compliance ISO 17034
Synonym Carbidopa, (2S)-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropanoic acid monohydrate
InChI Formula InChI=1S/C10H14N2O4.H2O/c1-10(12-11,9(15)16)5-6-2-3-7(13)8(14)4-6;/h2-4,12-14H,5,11H2,1H3,(H,15,16);1H2/t10-;/m0./s1
SMILES O.C[C@@](Cc1ccc(O)c(O)c1)(NN)C(=O)O
IUPAC Name (2S)-3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid;hydrate
Molecular Weight (g/mol) 244.24
Quantity 250 mg
Formula Weight 244.1059 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.