missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbic Anhydride, TRC
Specifications
Specifications
| Chemical Name or Material | Carbic Anhydride |
| CAS | 129-64-6 |
| Molecular Formula | C9H8O3 |
| Quantity | 25 g |
| Synonym | 5-Norbornene-endo-2,3-dicarboxylic anhydride, Bicyclo[2.2.1]-2-heptene-endo-5,6-dicarboxylic acid anhydride, Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid endo-cis-anhydride, Bicyclo[2.2.1]hept-5-ene-endo,endo-2,3-dicarboxylic anhydride, Bicyclo[2.2.1hept-5-ene-2-endo,3-endo-dicarboxylic anhydride, Carbic anhydride, Endic anhydride, Kayahard CD, NSC 102277, Nadic acid anhydride, Nadic anhydride, Norbornene-5,6-endo-dicarboxylic anhydride, cis-3,6-endo-Methylene-1,2,3,6-tetrahydrophthalic anhydride, cis-5-Norbornene-2,3-dicarboxylic anhydride, cis-5-Norbornene-endo-2,3-dicarboxylic acid anhydride, cis-5-Norbornene-endo-2,3-dicarboxylic anhydride, cis-endo-5-Norbornene-2,3-dicarboxylic acid anhydride, endo,cis-5-Norbornene-2,3-dicarboxylic acid anhydride, endo,cis-5-Norbornene-2,3-dicarboxylic anhydride, endo-3,6-Methylene-1,2,3,6-tetrahydrophthalic anhydride, endo-3,6-Methylene-4-cyclohexene-1,2-dicarboxylic anhydride, endo-5-Norbornene-2,3-dicarboxylic anhydride, endo-Bicyclo[2.2.1]hept-2-ene-5,6-dicarboxylic anhydride, endo-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride, endo-Carbic anhydride, endo-Himic acid anhydride, endo-Norbornene-2,3-dicarboxylic acid anhydride, endo-cis-3,6-endo-Methylene-delta4-tetrahydrophthalic anhydride, endo-cis-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid anhydride, endo-cis-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride |
| InChI Formula | InChI=1S/C9H8O3/c10-8-6-4-1-2-5(3-4)7(6)9(11)12-8/h1-2,4-7H,3H2/t4-,5+,6-,7+ |
| SMILES | O=C1OC(=O)[C@@H]2[C@H]3C[C@H](C=C3)[C@H]12 |
| IUPAC Name | 3a,4,7,7a-Tetrahydro-4,7-methanoisobenzofuran-1,3-dione |
| Molecular Weight (g/mol) | 164.16 |
| Formula Weight | 164.0473 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?