missing translation for 'onlineSavingsMsg'
Learn More

Carbic Anhydride, TRC

Product Code. 30359470
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
missing translation for 'unitSize'
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359470 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30359470 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCC176150100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbic Anhydride
CAS 129-64-6
Molecular Formula C9H8O3
Quantity 100 g
Synonym 5-Norbornene-endo-2,3-dicarboxylic anhydride, Bicyclo[2.2.1]-2-heptene-endo-5,6-dicarboxylic acid anhydride, Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid endo-cis-anhydride, Bicyclo[2.2.1]hept-5-ene-endo,endo-2,3-dicarboxylic anhydride, Bicyclo[2.2.1hept-5-ene-2-endo,3-endo-dicarboxylic anhydride, Carbic anhydride, Endic anhydride, Kayahard CD, NSC 102277, Nadic acid anhydride, Nadic anhydride, Norbornene-5,6-endo-dicarboxylic anhydride, cis-3,6-endo-Methylene-1,2,3,6-tetrahydrophthalic anhydride, cis-5-Norbornene-2,3-dicarboxylic anhydride, cis-5-Norbornene-endo-2,3-dicarboxylic acid anhydride, cis-5-Norbornene-endo-2,3-dicarboxylic anhydride, cis-endo-5-Norbornene-2,3-dicarboxylic acid anhydride, endo,cis-5-Norbornene-2,3-dicarboxylic acid anhydride, endo,cis-5-Norbornene-2,3-dicarboxylic anhydride, endo-3,6-Methylene-1,2,3,6-tetrahydrophthalic anhydride, endo-3,6-Methylene-4-cyclohexene-1,2-dicarboxylic anhydride, endo-5-Norbornene-2,3-dicarboxylic anhydride, endo-Bicyclo[2.2.1]hept-2-ene-5,6-dicarboxylic anhydride, endo-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride, endo-Carbic anhydride, endo-Himic acid anhydride, endo-Norbornene-2,3-dicarboxylic acid anhydride, endo-cis-3,6-endo-Methylene-delta4-tetrahydrophthalic anhydride, endo-cis-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid anhydride, endo-cis-Bicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic anhydride
InChI Formula InChI=1S/C9H8O3/c10-8-6-4-1-2-5(3-4)7(6)9(11)12-8/h1-2,4-7H,3H2/t4-,5+,6-,7+
SMILES O=C1OC(=O)[C@@H]2[C@H]3C[C@H](C=C3)[C@H]12
IUPAC Name 3a,4,7,7a-Tetrahydro-4,7-methanoisobenzofuran-1,3-dione
Molecular Weight (g/mol) 164.16
Formula Weight 164.0473
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.