missing translation for 'onlineSavingsMsg'
Learn More

Carbic anhydride, 99+%, Thermo Scientific Chemicals

Catalog Number 10006243
Click to view available options
Quantity:
100 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
129-64-6
C9H8O3
164.16
MFCD00151106,MFCD00069013,MFCD00630580,MFCD00151106
KNDQHSIWLOJIGP-UHFFFAOYNA-N
cis-5-norbornene-endo-2,3-dicarboxylic anhydride
126963424
4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
O=C1OC(=O)C2C3CC(C=C3)C12
This item is not returnable. View return policy
129-64-6
C9H8O3
164.16
MFCD00151106,MFCD00069013,MFCD00630580,MFCD00151106
KNDQHSIWLOJIGP-UHFFFAOYNA-N
cis-5-norbornene-endo-2,3-dicarboxylic anhydride
126963424
4-oxatricyclo[5.2.1.0²,⁶]dec-8-ene-3,5-dione
O=C1OC(=O)C2C3CC(C=C3)C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 162.0°C to 165.0°C
Color White
Density 1.0800g/mL
Infrared Spectrum Authentic
Assay Percent Range Balance: himic acid
Packaging Glass bottle
Refractive Index 1.4365
Quantity 100 g
Merck Index 15, 1797
Specific Gravity 1.08
Solubility Information Solubility in water: <0,01g/100g. Other solubilities: soluble in acetone,benzene,ethyl ether,ethanol,decomposes with alkalies
Formula Weight 164.16
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Carbic anhydride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'