missing translation for 'onlineSavingsMsg'
Learn More

(Carbethoxymethylene)triphenylphosphorane-13C2, TRC

Product Code. 30357854
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357854 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357854 Supplier Toronto Research Chemicals Supplier No. TRCC1759125MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (Carbethoxymethylene)triphenylphosphorane-13C2
CAS 90830-64-1
Molecular Formula C2013C2H21O2P
Quantity 5 mg
Synonym 2-(Triphenylphosphoranylidene)acetic Acid Ethyl Ester-13C2, (2-Ethoxy-2-oxoethylidene)triphenylphosphorane-13C2, Ethyl (triphenylphosphoranylidene)acetate-13C2, NSC 72406-13C2
InChI Formula InChI=1S/C22H21O2P/c1-2-24-22(23)18-25(19-12-6-3-7-13-19,20-14-8-4-9-15-20)21-16-10-5-11-17-21/h3-18H,2H2,1H3/i18+1,22+1
SMILES CCO[13C](=O)[13CH]=P(c1ccccc1)(c2ccccc2)c3ccccc3
IUPAC Name ethyl 2-(triphenyl-λ^{5}-phosphanylidene)acetate
Molecular Weight (g/mol) 350.36
Formula Weight 350.1346
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.