missing translation for 'onlineSavingsMsg'
Learn More

Carbetapentane citrate

Product Code. 15476389
Change view
Click to view available options
Quantity:
50 mg
1 g
Unit Size:
1g
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476389 50 mg 50mg
15466389 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476389 Supplier Thermo Scientific Alfa Aesar Supplier No. J64446.MF
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sigma receptor agonist | CAS: 23142-01-0 | C26H39NO10 | 525.595 g/mol

Sigma receptor agonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23142-01-0
Molecular Formula C26H39NO10
Molecular Weight (g/mol) 525.595
MDL Number MFCD00055697
InChI Key AKJDEXBCRLOVTH-UHFFFAOYSA-N
Synonym carbetapentane citrate, pentoxyverine citrate, loucarbate, pentoxiverine citrate, carbetapentane citrate salt, toclase, toclase citrate, carbetapentone citrate, unii-4sh0mfj5hj
PubChem CID 90010
IUPAC Name 2-[2-(diethylamino)ethoxy]ethyl 1-phenylcyclopentane-1-carboxylate;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES CCN(CC)CCOCCOC(=O)C1(CCCC1)C2=CC=CC=C2.C(C(=O)O)C(CC(=O)O)(C(=O)O)O

Specifications

Color White
Quantity 50 mg
Merck Index 14,1794
Formula Weight 633.7
Physical Form Powder
Chemical Name or Material Carbetapentane citrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.