missing translation for 'onlineSavingsMsg'
Learn More

Carbetapentane citrate, MedChemExpress

Product Code. 16452246
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16452246 500 mg 500mg
16408586 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16452246 Supplier Medchem Express Supplier No. HYB1055
 Restricted Item
Estimated Shipment: 30-09-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress Carbetapentane citrate is a selective inhibitor of the cough, with mild atropine-like effect and local anesthesia effect.

Carbetapentane citrate is a selective inhibitor of the cough, with mild atropine-like effect and local anesthesia effect.

Specifikationer

Chemical Name or Material Carbetapentane citrate
CAS 23142-01-0
Color White
Purity Grade Notes Research
Molecular Formula C26H39NO10
Quantity 500 mg
Synonym Pentoxyverine citrate
Solubility Information DMSO : ≥ 36 mg/mL (68.49 mM)
SMILES O=C(C1(C2=CC=CC=C2)CCCC1)OCCOCCN(CC)CC.O=C(CC(C(O)=O)(O)CC(O)=O)O
Molecular Weight (g/mol) 525.59
Formula Weight 525.59
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Visa mer Visa mindre

Research purposes only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.