missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Carbenicillin disodium salt, 90%

Product Code. p-7077546 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15525146 1 g 1g
15535146 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15525146 Supplier Thermo Scientific Chemicals Supplier No. 455360010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carbenicillin disodium salt, CAS # 4800-94-6, is the disodium salt form of carbenicillin, a semisynthetic compound with broad-spectrum antibacterial and β-lactamase-resistant activities.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Carbenicillin disodium salt is a semisynthetic compound with antibacterial and β-lactamase-resistant activities and is effective against gram-negative bacteria
  • In in vitro studies, carbenicillin disodium salt can totally suppress the proliferation of Agrobacterium

Application

  • Due to its antibacterial activity this compound can be used to control bacterial cell wall synthesis by inactivating transpeptidases on the inner surface of the bacterial cell membrane. This can lead to incomplete bacterial cell wall synthesis to eventually cause cell lysis
  • It has also been used in studies to evaluate the role of penicillin-sensitive transpeptidases in cell wall biosynthesis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4800-94-6
Molecular Formula C17H16N2Na2O6S
Molecular Weight (g/mol) 422.36
MDL Number MFCD00077683
InChI Key RTYJTGSCYUUYAL-YCAHSCEMSA-L
PubChem CID 20933
ChEBI CHEBI:34609
IUPAC Name disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
SMILES [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C([O-])=O)C3=CC=CC=C3)C(=O)N2[C@H]1C([O-])=O

Specifications

Type Carbenicillin Disodium Salt
Melting Point >200.0°C
Infrared Spectrum Authentic
Water 6% max.
Quantity 1 g
Flash Point 400°C
Boiling Point 738.0°C
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and methanol
Formula Weight 422.36
Percent Purity 90%
Chemical Name or Material Carbenicillin disodium salt
Show More Show Less
compliance-icons
Product Identifier
  • Carbenicillin disodium salt
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.