missing translation for 'onlineSavingsMsg'
Learn More

Carbenicillin disodium, MedChemExpress

Product Code. 16422219
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16422219 1 g 1g
16440978 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16422219 Supplier Medchem Express Supplier No. HYB0525A
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Carbenicillin disodium is a beta-lactam penicillin derivative that interference with final stage of bacterial cell wall synthesis.

Carbenicillin disodium is a beta-lactam penicillin derivative that interference with final stage of bacterial cell wall synthesis.

Specifications

Chemical Name or Material Carbenicillin disodium
CAS 4800-94-6
Color White
Purity Grade Notes Research
Molecular Formula C17H16N2Na2O6S
Quantity 1 g
Synonym Sodium carbenicillin
Solubility Information H2O : 125 mg/mL (295.96 mM; Need ultrasonic) ∣DMSO : 62.5 mg/mL (147.98 mM; Need ultrasonic)
SMILES O=C([C@@H](C(C)(C)S[C@]1([H])[C@@H]2NC(C(C(O[Na])=O)C3=CC=CC=C3)=O)N1C2=O)O[Na]
Molecular Weight (g/mol) 422.36
Formula Weight 422.36
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.