missing translation for 'onlineSavingsMsg'
Learn More

Carbenicillin Disodium (90%), TRC

Product Code. 30353885
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30353885 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30353885 Supplier Toronto Research Chemicals Supplier No. TRCC1760805G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbenicillin disodium
CAS 4800-94-6
Molecular Formula C17 H16 N2 O6 S . 2 Na
Quantity 5 g
Synonym 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[(2-carboxy-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-, sodium salt (1:2), (2S,5R,6R)-, 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[(carboxyphenylacetyl)amino]-3,3-dimethyl-7-oxo-, disodium salt, (2S,5R,6R)- (9CI), 4-Thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 6-[(carboxyphenylacetyl)amino]-3,3-dimethyl-7-oxo-, disodium salt, [2S-(2α,5α,6β)]-, Malonamic acid, N-(2-carboxy-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]hept-6-yl)-2-phenyl-, disodium salt (7CI,8CI), Anabactyl, BRL 2064, CP 15639-2, Carbapen, Carbecin, Carbenicillin disodium, Carbenicillin disodium salt, Carbenicillin sodium, Carbenicilline disodium, Carboxybenzylpenicillin sodium, Disodium (α-carboxybenzyl)penicillin, Disodium carbenicillin, Fugacillin, Geocillin, Geopen, Gripenin, Hyoper, Microcillin, NSC 111071, Piopen, Pyopen, Pyopene, Sodium carbenicillin, α-Carboxybenzylpenicillin disodium salt, α-Carboxybenzylpenicillin sodium salt
InChI Formula InChI=1S/C17H18N2O6S.2Na/c1-17(2)11(16(24)25)19-13(21)10(14(19)26-17)18-12(20)9(15(22)23)8-6-4-3-5-7-8;;/h3-7,9-11,14H,1-2H3,(H,18,20)(H,22,23)(H,24,25);;/q;2*+1/p-2/t9?,10-,11+,14-;;/m1../s1
SMILES [Na+].[Na+].CC1(C)S[C@@H]2[C@H](NC(=O)C(C(=O)[O-])c3ccccc3)C(=O)N2[C@H]1C(=O)[O-]
IUPAC Name disodium;(2S,5R,6R)-6-[(2-carboxylato-2-phenylacetyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Molecular Weight (g/mol) 422.36
Formula Weight 422.0524
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.