missing translation for 'onlineSavingsMsg'
Learn More

Melford Carbenicillin

Product Code. 30018167
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30018167 5 g 5g
30018164 1 g 1g
30018165 100 g 100g
30018166 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 30018167 Supplier Melford Supplier No. C460005.0
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Carbenicillin, Disodium Salt is an antibiotic that belongs to the penicillin class of antibiotics and is effective against both Gram-positive and Gram-negative bacteria.

Carbenicillin is a semisynthetic derivative of penicillin and is known for its stability in aqueous solutions.

In the field of molecular biology and genetic engineering, Carbenicillin, Disodium Salt is commonly used as a selective agent in plasmid-based cloning experiments. It functions by inhibiting bacterial cell wall synthesis, leading to cell death.

Researchers often introduce a plasmid containing a resistance gene for Carbenicillin into host cells. The cells that successfully take up the plasmid and express the resistance gene can survive in the presence of Carbenicillin, while non-transformed cells are eliminated.

Carbenicillin, Disodium Salt is also used in microbiology and cell culture applications. It is effective in preventing the growth of bacterial contaminants in cell cultures, ensuring the purity of cell lines.

Additionally, it is employed for the isolation and maintenance of strains with known or engineered resistance to Carbenicillin.

Specifications

Chemical Name or Material Carbenicillin
CAS 4800-94-6
Quantity 5 g
Molecular Formula C17H16N2Na2O6S
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.