missing translation for 'onlineSavingsMsg'
Learn More

Carbendazim, TRC

Product Code. 30134277
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134277 1 g 1g
30040546 100 mg 100mg
17927149 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134277 Supplier Toronto Research Chemicals Supplier No. TRCC1759001G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10605-21-7
Molecular Formula C9 H9 N3 O2
Molecular Weight (g/mol) 191.19
Synonym 2-Benzimidazolecarbamic acid, methyl ester (6CI,7CI,8CI), Carbamic acid, 1H-benzimidazol-2-yl-, methyl ester (9CI), 2-(Carbomethoxyamino)benzimidazole, 2-(Methoxycarbamoyl)benzimidazole, 2-(Methoxycarboxamido)benzimidazole, 2-[(Methoxycarbonyl)amino]benzimidazole, A 118, A 118 (pesticide), Agrizim, Aimcozim, Amolden TSM 32, Antibac MF, B 350, BAS 3460, BAS 3460F, BAS 67054F, BCM, BCM (fungicide), BMK, BMK (fungicide), Battal, Bavistan, Bavistin, Bavistin 25SD, Bavistin 3460, Bavistin 50SD, Bavistin 50WP, Bavistin DF, Bavistin FL, Bavistine, Bendazim, Benfil, Bengard, Bercema-Bitosen, Biocut BM 10, Bitosen, CTR 6669, Carbate, Carben VL, Carbendazim, Carbendazime, Carbendazol, Carbendazole, Carbendor, Carbex 500SC, Carbomax, Cekudazim, Corbel, Corzim, Corzim 50WP, Custos, Defensor, Delsene, Delsene 10, Derosal, Derosal 60PM, Derosol 60WP, Derroprene, Dhanustin, EK 578, Elsa, Equitdazin, FB 642, Falicarben, Ferazim, Funaben, Funaben 3, Funaben 50, Fungoxan, G 665, GLSTIN, Galofungin, Garbenda, HOE 17411, IPO 1250, IPO Y, Jkstein, Karben, Karben Stefes Flo, Karben flo Stefes, Kemdazin, Kolfugo, Kolfugo 25FW, Kolfugo Extra, Kolfugo Super, Lignasan, MBC, Mecarzole, Medamine, Methoxycarbonylaminobenzimidazole, Methyl 1H-2-benzimidazolecarbaminate, Methyl 1H-benzimidazol-2-ylcarbamate, Methyl 1H-benzimidazole-2-carbamate, Methyl 2-benzimidazolecarbamate, Methyl 2-benzimidazolylcarbamate, Methyl benzimidazolylcarbamate, Myco, NSC 109874, Olgin, Olgin (fungicide), Pillarstin, Preparation G 665, Preventol BCM, Protek, San-ai 700, Sarfun, Spin, Spin (pesticide), Spin Flo, Stein, Stempor, Subeej, Supercarb, Supercarb (fungicide), Thicoper, Triticol, Troysan CF 20, U 32104, Zhiweiling
IUPAC Name methyl N-(1H-benzimidazol-2-yl)carbamate
SMILES COC(=O)Nc1nc2ccccc2[nH]1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H9N3O2/c1-14-9(13)12-8-10-6-4-2-3-5-7(6)11-8/h2-5H,1H3,(H2,10,11,12,13)
Formula Weight 191.0695
Percent Purity >95%
Chemical Name or Material Carbendazim
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.