missing translation for 'onlineSavingsMsg'
Learn More

Carbendazim, MedChemExpress

Product Code. 16466236
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
Unit Size:
100mg
1g
500mg
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16466236 100 mg 100mg
16471359 1 g 1g
16451676 500 mg 500mg
16428447 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16466236 Supplier Medchem Express Supplier No. HY13582
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Carbendazim is a potent and orally active broad-spectrum benzimidazole fungicide and can be acts as a pesticide for fungal diseases research, such as Seproria, Fusarium and Sclerotina. Carbendazim is a benzimidazole (HY-Y1825) derivative with antitumor activity and used for cancer research, especially advanced solid tumors and lymphoma.

Carbendazim is a potent and orally active broad-spectrum benzimidazole fungicide and can be acts as a pesticide for fungal diseases research, such as Seproria, Fusarium and Sclerotina. Carbendazim is a benzimidazole (HY-Y1825) derivative with antitumor activity and used for cancer research, especially advanced solid tumors and lymphoma.

Specifications

Chemical Name or Material Carbendazim
Physical Form Solid
CAS 10605-21-7
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H9N3O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 6.8 mg/mL (35.57 mM; Need ultrasonic)
SMILES O=C(OC)NC1=NC2=CC=CC=C2N1
Molecular Weight (g/mol) 191.19
Formula Weight 191.19
Percent Purity 99.81%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.