missing translation for 'onlineSavingsMsg'
Learn More

Carbazole, 95%, Thermo Scientific Chemicals

Catalog Number 11344569
Click to view available options
Quantity:
50 g
100 g
500 g
This item is not returnable. View return policy
86-74-8
C12H9N
167.21
MFCD00004960
UJOBWOGCFQCDNV-UHFFFAOYSA-N
carbazole, dibenzopyrrole, diphenylenimine, 9-azafluorene, diphenylenimide, diphenyleneimine, dibenzo b,d pyrrole, usaf ek-600, unii-0p2197hhhn
6854
CHEBI:27543
9H-carbazole
N1C2=C(C=CC=C2)C2=C1C=CC=C2
This item is not returnable. View return policy
86-74-8
C12H9N
167.21
MFCD00004960
UJOBWOGCFQCDNV-UHFFFAOYSA-N
carbazole, dibenzopyrrole, diphenylenimine, 9-azafluorene, diphenylenimide, diphenyleneimine, dibenzo b,d pyrrole, usaf ek-600, unii-0p2197hhhn
6854
CHEBI:27543
9H-carbazole
N1C2=C(C=CC=C2)C2=C1C=CC=C2

Carbazole and its derivatives are widely used as an intermediate in synthesis of pharmaceuticals, agrochemicals, dyes, pigments and other organic compounds. carbazole is also used in luminescence chemistry as a photosensitizing and additional charge transport material. Carbazole structure is a motif in pharmaceuticals such as carvedilol used to treat high blood pressure and to prevent cardiac arrhythmias and angina.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Carbazole and its derivatives are widely used as an intermediate in synthesis of pharmaceuticals, agrochemicals, dyes, pigments and other organic compounds. carbazole is also used in luminescence chemistry as a photosensitizing and additional charge transport material. Carbazole structure is a motif in pharmaceuticals such as carvedilol used to treat high blood pressure and to prevent cardiac arrhythmias and angina.

Solubility
It is insoluble in water.

Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point 240°C to 246°C
Density 1.15
Boiling Point 354°C to 356°C
Flash Point 220°C (428°F)
Odor Odorless
Quantity 50 g
UN Number UN3077
Beilstein 3956
Merck Index 14,1790
Solubility Information It is insoluble in water.
Formula Weight 167.21
Percent Purity 95%
Chemical Name or Material Carbazole
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'