missing translation for 'onlineSavingsMsg'
Learn More

Carbazochrome sodium sulfonate, MedChemExpress

Product Code. 16478336
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16478336 100 mg 100mg
16434377 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478336 Supplier Medchem Express Supplier No. HYB0491A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Carbazochrome sodium sulfonate (AC-17) is a capillary stabiliser and used for the research of haemorrhage. Carbazochrome sodium sulfonate is an antihemorrhagic agent.

Carbazochrome sodium sulfonate (AC-17) is a capillary stabiliser and used for the research of haemorrhage. Carbazochrome sodium sulfonate is an antihemorrhagic agent.

Specifications

Chemical Name or Material Carbazochrome sodium sulfonate
CAS 51460-26-5
Color Orange
Purity Grade Notes Research
Molecular Formula C10H11N4NaO5S
Quantity 100 mg
Synonym AC-17
Solubility Information DMSO : ≥ 49 mg/mL (152.05 mM)
SMILES O=S(C(CC1=C/C2=N/NC(N)=O)N(C)C1=CC2=O)(O[Na])=O
Molecular Weight (g/mol) 322.27
Formula Weight 322.27
Percent Purity 98.69%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.