missing translation for 'onlineSavingsMsg'
Learn More

Carbazochrome Sodium Sulfonate Hydrate, TRC

Product Code. 30367302
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30367302 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367302 Supplier Toronto Research Chemicals Supplier No. TRCC1761901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbazochrome Sodium Sulfonate Hydrate
CAS 51460-26-5
Molecular Formula C10 H11 N4 O5 S . Na . 3 H2 O
Quantity 1 g
Synonym 5-[2-(Aminocarbonyl)hydrazinylidene]-2,3,5,6-tetrahydro-1-methyl-6-oxo-1H-indole-2-Sulfonic Acid Sodium Salt Hydrate, Adenaron Hydrate, Adona Hydrate, Adrenochrome sulfonate Ac 17 Hydrate, Carbazon Hydrate, Donaseven Hydrate, Emex Hydrate, Odanon Hydrate, Tazin Hydrate
InChI Formula InChI=1S/C10H12N4O5S.Na.3H2O/c1-14-7-4-8(15)6(12-13-10(11)16)2-5(7)3-9(14)20(17,18)19;;;;/h2,4,9H,3H2,1H3,(H3,11,13,16)(H,17,18,19);;3*1H2/q;+1;;;/p-1/b12-6+;;;;
SMILES O.O.O.[Na+].CN1C(CC2=C\C(=N/NC(=O)N)\C(=O)C=C12)S(=O)(=O)[O-]
IUPAC Name sodium;(5E)-5-(carbamoylhydrazinylidene)-1-methyl-6-oxo-2,3-dihydroindole-2-sulfonate;trihydrate
Molecular Weight (g/mol) 376.32
Formula Weight 376.0665
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.