missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbazochrome Sodium Sulfonate Hydrate, TRC
Specifications
Specifications
| Chemical Name or Material | Carbazochrome Sodium Sulfonate Hydrate |
| CAS | 51460-26-5 |
| Molecular Formula | C10 H11 N4 O5 S . Na . 3 H2 O |
| Quantity | 50 mg |
| Synonym | 5-[2-(Aminocarbonyl)hydrazinylidene]-2,3,5,6-tetrahydro-1-methyl-6-oxo-1H-indole-2-Sulfonic Acid Sodium Salt Hydrate, Adenaron Hydrate, Adona Hydrate, Adrenochrome sulfonate Ac 17 Hydrate, Carbazon Hydrate, Donaseven Hydrate, Emex Hydrate, Odanon Hydrate, Tazin Hydrate |
| InChI Formula | InChI=1S/C10H12N4O5S.Na.3H2O/c1-14-7-4-8(15)6(12-13-10(11)16)2-5(7)3-9(14)20(17,18)19;;;;/h2,4,9H,3H2,1H3,(H3,11,13,16)(H,17,18,19);;3*1H2/q;+1;;;/p-1/b12-6+;;;; |
| SMILES | O.O.O.[Na+].CN1C(CC2=C\C(=N/NC(=O)N)\C(=O)C=C12)S(=O)(=O)[O-] |
| IUPAC Name | sodium;(5E)-5-(carbamoylhydrazinylidene)-1-methyl-6-oxo-2,3-dihydroindole-2-sulfonate;trihydrate |
| Molecular Weight (g/mol) | 376.32 |
| Formula Weight | 376.0665 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?