missing translation for 'onlineSavingsMsg'
Learn More

Carbasalate Calcium, TRC

Product Code. 30362027
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30362027 500 mg 500mg
30355764 1 g 1g
30354834 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30362027 Supplier Toronto Research Chemicals Supplier No. TRCC176820500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5749-67-7
Molecular Formula 2(C9H7O4) . CH4N2O . Ca
Molecular Weight (g/mol) 458.4
Synonym Calpirinsan, Calcium acetyl salicylate carbamide, Iromin, Omegin, Solupsan, SoluspanUrea calcium acetylsalicylate, Ursinus, Salicylic acid acetate, calcium salt, compd. with urea, Acetylsalicylic acid calcium salt complex with urea
IUPAC Name calcium;2-acetyloxybenzoate;urea
SMILES [Ca+2].CC(=O)Oc1ccccc1C(=O)[O-].CC(=O)Oc2ccccc2C(=O)[O-].NC(=O)N

Specifications

Quantity 500 mg
InChI Formula InChI=1S/2C9H8O4.CH4N2O.Ca/c2*1-6(10)13-8-5-3-2-4-7(8)9(11)12;2-1(3)4;/h2*2-5H,1H3,(H,11,12);(H4,2,3,4);/q;;;+2/p-2
Formula Weight 458.0638
Chemical Name or Material Carbasalate Calcium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.