missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Carbaryl Monoclonal Antibody (4H3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549726
This item is not returnable.
View return policy
Description
Antibody tested in indirect ELISA.
Carbaryl is an insecticide that belongs to the chemical class of carbamates. It is commonly used in agriculture to control pests in crops, such as fruits, vegetables, and cereals. Carbaryl works by disrupting the nervous system of insects, causing paralysis and ultimately death. It is also used in public health programs to control disease-carrying insects, such as mosquitoes and ticks.
Specifications
| Carbaryl | |
| Monoclonal | |
| 1.42 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Conjugate of CBR hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
| 4H3 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction