missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbamylcholine Chloride, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | Carbachol |
| CAS | 51-83-2 |
| Molecular Formula | C6 H15 N2 O2 . Cl |
| Quantity | 10 g |
| Synonym | Carbachol, 2-(Carbamoyloxy)-N,N,N-trimethylethanaminium chloride, Choline, chloride, carbamate (6CI,8CI), Ethanaminium, 2-[(aminocarbonyl)oxy]-N,N,N-trimethyl-, chloride (9CI), (2-Carbamoyloxyethyl)trimethylammonium chloride, (2-Hydroxyethyl)trimethyl ammonium chloride carbamate, Carbachol chloride, Carbacholin, Carbacholine, Carbacholine chloride, Carbacolina, Carbaminocholine chloride, Carbaminoylcholine chloride, Carbamiotin, Carbamoylcholine chloride, Carbamoylcholine-hydrochloride, Carbamylcholine chloride, Carbastat Intraocular, Carbochol, Carbocholin, Carbocholine, Carbyl, Carcholin, Choline carbamate chloride, Coletyl, Doryl, Doryl (pharmaceutical), Isopto Carbachol, Jestryl, Lentin, Miostat, Moryl, Vasoperif, Ethanaminium, 2-[(aminocarbonyl)oxy]-N,N,N-trimethyl-, chloride (1:1), gamma-Carbamoyl choline chloride |
| InChI Formula | InChI=1S/C6H14N2O2.ClH/c1-8(2,3)4-5-10-6(7)9;/h4-5H2,1-3H3,(H-,7,9);1H |
| SMILES | [Cl-].C[N+](C)(C)CCOC(=O)N |
| IUPAC Name | 2-carbamoyloxyethyl(trimethyl)azanium;chloride |
| Molecular Weight (g/mol) | 182.65 |
| Formula Weight | 182.0822 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction