missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbamyl Benzyl Ester, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Carbamyl Benzyl Ester |
| CAS | 621-84-1 |
| Molecular Formula | C8H9NO2 |
| Quantity | 25 g |
| Synonym | Carbamic Acid Phenylmethyl Ester, Benzyl Aminoformate, Benzyl Carbamate, NSC 25317, O-Benzylcarbamate |
| InChI Formula | InChI=1S/C8H9NO2/c9-8(10)11-6-7-4-2-1-3-5-7/h1-5H,6H2,(H2,9,10) |
| SMILES | NC(=O)OCc1ccccc1 |
| IUPAC Name | benzyl carbamate |
| Molecular Weight (g/mol) | 151.16 |
| Formula Weight | 151.0633 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction