missing translation for 'onlineSavingsMsg'
Learn More

Carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy, free radical, 99%

Product Code. 11375079
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11375079 1 g 1g
11385079 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11375079 Supplier Thermo Scientific Alfa Aesar Supplier No. A17381.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3229-73-0 | C9H16N2O2 | 184.24 g/mol

It is employed as a highly reactive spin-label.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a highly reactive spin-label.

Solubility
Soluble in acetone, chloroform, dichloromethane, ethyl acetate and methanol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3229-73-0
Molecular Formula C9H16N2O2
Molecular Weight (g/mol) 184.24
MDL Number MFCD00005341
InChI Key GMXOTWPDUBXCMS-UHFFFAOYSA-N
Synonym 2,2,5,5-Tetramethyl-3-pyrrolin-1-oxyl-3-carboxamide
IUPAC Name 1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxamide
SMILES CC1(C)C=C(C(N)=O)C(C)(C)N1O

Specifications

Melting Point 198°C to 200°C
Quantity 1 g
Beilstein 4138020
Solubility Information Soluble in acetone,chloroform,dichloromethane,ethyl acetate and methanol.
Formula Weight 183.23
Percent Purity 99%
Chemical Name or Material Carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy, free radical

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.